5-Aminoimidazole-4-carboxamide 1-beta-D-Ribofuranoside ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
AICAR, 5-Amino-1-(β-D-ribofuranosyl)-1H-imidazole-4-carboxamide, Acadesine
AG-CR1-0061-M010
AG-CR1-0061-M050
AG-CR1-0061-M100
102516-100EA
34.62
USD
102516-100
102516-102
102988-532
5-Aminoimidazole-4-carboxamide 1-beta-D-Ribofuranoside ≥98% (by HPLC)
5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside
Cell permeable AMP-activated protein kinase (AMPK) activator. Insulin mimetic. Adipocyte differentiation inhibitor. Apoptosis inducer. PPARalpha inhibitor. mTOR inhibitor. P70S6K inhibitor. LPS-induced TNF-alpha production inhibitor. TORC2 phosphorylation inducer. Anti-inflammatory. Anti-tumor compound. Autophagy inhibitor. HSP90 inhibitor. Autophagy inducer
Formula:
C₉H₁₄N₄O₅ MW: 258.23 g/mol |
MDL Number:
MFCD00869751 CAS Number: 2627-69-2 |
Specification Test Results
Purity | ≥98% (HPLC) |
Identity | Determined by 1H-NMR. |
Appearance | Off-white solid. |
Solubility | Soluble in water (9 mg/ml), DMSO or dimethyl formamide |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...