Celastrol

Supplier: ADIPOGEN CORP MS

Synonyms: Tripterin

AG-CN2-0460-M005 AG-CN2-0460-M010 AG-CN2-0460-M050
102988-348EA 83.08 USD
102988-348 102988-350 102988-352
Celastrol
Celastrol

Anti-obesity agent. Potent leptin sensitizer. Antioxidant. Potent lipid peroxidation inhibitor. NADPH oxidases (NOX1, 2, 4 and 5) inhibitor. Anti-inflammatory. NF-kappaB inhibitor. HSP90 inhibitor and HSP70 inducer. Anticancer agent. Apoptosis and autophagy inducer. Inhibits PI3K/Akt signaling pathways. Anti-metastatic and anti-angiogenic compound. Reduces VEGFR expression and inhibits HIF-1alpha pathway. Topoisomerase II inhibitor. 20S proteasome chymotrypsin-like activity inhibitor. Potential antiviral and anti-Alzheimer's agent.

Formula: C₂₉H₃₈O₄
MW: 450.62 g/mol
MDL Number: MFCD03424073
CAS Number: 34157-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR