Isoliquiritigenin

Supplier: ADIPOGEN CORP MS

Synonyms: (E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one, 2',4,4'-Trihydroxychalcone, (E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)-2-propen-1-one

AG-CN2-0459-M010 AG-CN2-0459-M050
102988-344EA 166.15 USD
102988-344 102988-346
Isoliquiritigenin
Isoliquiritigenin

Anticancer. Apoptosis inducer and cell proliferation inhibitor. Autophagy inducer. Anti-angiogenic. VEGF/VEGFR-2 signaling inhibitor. Antioxidant and antibacterial. AMPK-mediated GSK3beta inhibitor. Anti-inflammatory. Inhibits translocation and activation of NF-kappaB by blocking phosphorylation and subsequent degradation of IkappaBalpha. Downregulates TRIF-dependent signaling pathways of TLRs by inhibiting TBK1 kinase activity. Blocks LPS-induced TLR4/MD2 complex signaling. NLRP3 inflammasome activation inhibitor. Inhibits NLRP3-activated ASC oligomerization. Can block the NLRP3 inflammasome at both the priming step and the activation step. H(2) histamine receptor signaling inhibitor. Anti-diabetic. Antihyperglycemic. Aldose reductase inhibitor. Vasorelaxant. Soluble guanylyl cyclase activator. GABAA receptor positive allosteric modulator. Potent cystic fibrosis transmembrane conductance regulator (CFTR) inhibitor. Shown to inhibit breast cancer stem cells through WIF1 demethylation. Shows outstanding matrix properties for MALDI-TOF-MS analysis of oligosaccharides.

Formula: C₁₅H₁₂O₄
MW: 256.26 g/mol
MDL Number: MFCD00075907
CAS Number: 961-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR