(1S,3R)-(-)-Camphoric acid
Supplier: ADIPOGEN CORP MS
Synonyms:
(1S,3R)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid
AG-CN2-0456-M500
AG-CN2-0456-G001
AG-CN2-0456-GG25
102988-330EA
103.85
USD
102988-330
102988-332
102988-334
(1S,3R)-(-)-Camphoric acid
(1S,3R)-(-)-Camphoric acid
Chirality inducing agent. Useful tool as resolving agent to separate the isomers and purify racemic mixtures (e.g. DL-lysine) by fractionally crystallizing the product to obtain the pure isomer (e.g. D-lysine). Can also be used as building block.
Formula:
C₁₀H₁₆O₄ MW: 200.23 g/mol |
MDL Number:
MFCD00064937 CAS Number: 560-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...