Triptolide

Supplier: ADIPOGEN CORP MS

AG-CN2-0448-M001 AG-CN2-0448-M005 AG-CN2-0448-M010
102988-300EA 34.62 USD
102988-300 102988-302 102988-304
Triptolide
Triptolide

Potent immunosuppressant with anti-inflammatory and antitumor properties. Exhibits potent antiproliferative activity, induces apoptosis, modulates autophagy and inhibits cell cycle regulators. Inhibits global gene transcription by inducing degradation of RNA polymerase II (Pol II) and by inhibiting the ATPase activity of XPB (a subunit of the general transcription factor TFIIH). Human dCTP pyrophosphatase 1 inhibitor. Interferes with a number of transcription factors including p53, NF-kappaB, nuclear factor of activated T cells (NFAT) and heat shock factor protein 1 (HSF-1). Shown to inhibit aromatase activity. Shows antifertility activity and antipolycystic kidney disease effects.

Formula: C₂₀H₂₄O₆
MW: 360.41 g/mol
MDL Number: MFCD00210565
CAS Number: 38748-32-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR