Itaconic acid

Supplier: ADIPOGEN CORP MS

Synonyms: 2-Methylenesuccinic acid, Methylenesuccinic acid

AG-CN2-0426-G001 AG-CN2-0426-G005
102988-238EA 27.69 USD
102988-238 102988-240
Itaconic acid
Itaconic acid

Mammalian antibacterial metabolite. Inhibitor of isocitrate lyase, the key enzyme of the glyoxylate cycle, essential for bacterial growth. Antimicrobial. Antibacterial. Inhibits the growth of bacteria expressing isocitrate lyase. Phosphofructokinase (PFKII) inhibitor. Suppresses glycolysis by decreasing the level of fructose 2,6-bisphosphate, resulting in decreased visceral fat. Might be involved in the regulation of metabolism. One of the most highly induced metabolites in activated macrophages. Exerts anti-inflammatory effects in vitro and in vivo. Regulates succinate levels by inhibiting succinate dehydrogenase (SDH). Regulates mitochondrial respiration and cytokine production. Might induce inflammasome activation and consequently IL-1beta secretion by generating ROS.

Formula: C₅H₆O₄
MW: 130.1 g/mol
Boiling Pt: 381
MDL Number: MFCD00004260
CAS Number: 97-65-4
Merck Index: 13,05264

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR