Papaverine hydrochloride
Supplier: ADIPOGEN CORP MS
AG-CN2-0414-G001
AG-CN2-0414-G005
102988-184EA
27.69
USD
102988-184
102988-186
Papaverine hydrochloride
Papaverine hydrochloride
Smooth muscle relaxant and vasodilator. Phosphodiesterase PDE10A inhibitor. Antiviral. Antipsychotic. Potential inducer of browning in WAT.
Formula:
C₂₀H₂₁NO₄·HCl MW: 375.85 g/mol |
MDL Number:
MFCD00012745 CAS Number: 61-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...