Trichostatin A

Supplier: ADIPOGEN CORP MS

Synonyms: TSA, (R,2E,4E)-7-(4-(dimethylamino)phenyl)-N-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide, [R-(E,E)]-7-[4-(Dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide

AG-CN2-0108-M001 AG-CN2-0108-M005
102987-374EA 124.62 USD
102987-374 102987-376
Trichostatin A
Trichostatin A

Antibiotic. Antifungal. Antiprotozoal. Potent and reversible, cell permeable inhibitor of histone deacetylase (HDAC). Immunosuppressive. Anticancer compound. Antifibrogenic. Apoptosis inducer. Induces cell growth arrest at both G1 and G2/M phases. Enhances the efficacy of anticancer agents that target DNA. Inactivates mitotic spindle checkpoint. Smooth muscle cell proliferation inhibitor. Telomerase inhibitor. Downregulates DNA methyltransferase DNMT1 and affects DNA methylation. Anti-inflammatory. Inhibits osteoclastogenesis and bone resorption. Promotes either self-renewal or differentiation of embryonic stem cells.

Formula: C₁₇H₂₂N₂O₃
MW: 302.37 g/mol
MDL Number: MFCD03848392
CAS Number: 58880-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR