Luteolin

Supplier: ADIPOGEN CORP MS

Synonyms: 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4-benzopyrone, Natural Yellow 2, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4H-chromen-4-one, 3',4',5,7-Tetrahydroxyflavone

AG-CN2-0098-M005 AG-CN2-0098-M025
102987-330EA 20.77 USD
102987-330 102987-332
Luteolin
Luteolin

Anticancer and antimetastatic. alpha-Glucosidase inhibitor. Topoisomerase I Inhibitor. Apoptosis inducer. Anti-inflammatory. 15-Lipoxygenase (15-LOX) inhibitor. IL-6 production inhibitor. Fatty acid synthase (FAS) inhibitor. Antioxidant. Free radical scavenger. Antiadipogenic. PPAR-gamma inhibitor. Neuroprotective. Monoamine transporter activator. Inhibitor of phosphodiesterases 1-5 (PDE1-5). PKC(epsilon) and Src kinase inhibitor. HDAC inhibitor. Autophagy modulator Phosphatidylinositol 3-kinase (PI3K) / Akt signaling inhibitor. p90 ribosomal S6 kinase (RSK1/RSK2) kinase activity inhibitor.

Formula: C₁₅H₁₀O₆
MW: 286.24 g/mol
MDL Number: MFCD00017309
CAS Number: 491-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR