Piceatannol

Supplier: ADIPOGEN CORP MS

Synonyms: (E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol

AG-CN2-0086-M001 AG-CN2-0086-M005 AG-CN2-0086-M025
102987-292EA 41.54 USD
102987-292 102987-294 102987-296
Piceatannol
Piceatannol

Wide range of tyrosine and serine/threonine protein kinase inhibitor, including Syk, p56lck, PKA, PKC, MLCK, CDPK, JNK and PI3K. Inhibits the tyrosine phosphorylation of STAT3 and STAT5. Potent apoptosis inducer. Potent anticancer compound. Suppresses NF-kB activation through IkBalpha kinase inhibition. Activator of human deacetylase SIRT1 (sirtuin 1). Potent antioxidant with anti-proliferative, anti-inflammatory and cardioprotective properties. Neuoprotective. Adipogenesis inhibitor. Promotes glucose uptake, AMPK phosphorylation and GLUT4 translocation. Autophagy inducer

Formula: C₁₄H₁₂O₄
MW: 244.2427 g/mol
MDL Number: MFCD00221715
CAS Number: 10083-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR