Imperatorin
Supplier: ADIPOGEN CORP MS
Synonyms:
9-((3-Methylbut-2-en-1-yl)oxy)-7H-furo[3,2-g]chromen-7-one
AG-CN2-0084-M001
AG-CN2-0084-M005
102987-286EA
48.46
USD
102987-286
102987-288
Imperatorin
Imperatorin
Anticancer compound. Apoptosis inducer. Induces DNA fragmentation at the G1/S phase of the cell cycle. HIV-1 replication inhibitor. Anti-inflammatory. Vasodilatation inducer. Increases BMP-2 expression (mRNA) and enhance bone formation. Modulator of SMAD1/5/8, p38, ERK pathways. Exerts anti-hypertrophic effects. Caspase-1 activity inhibitor in vivo and in vitro. Antibacterial and antilisteria compound. L-Type calcium channel blocker.
Formula:
C₁₆H₁₄O₄ MW: 270.284 g/mol |
MDL Number:
MFCD00016881 CAS Number: 482-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...