Gentamicin sulphate USP

Supplier: ADIPOGEN CORP MS

Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

AG-CN2-0066-G001 AG-CN2-0066-G005 AG-CN2-0066-G025
102987-214EA 48.46 USD
102987-214 102987-216 102987-218
Gentamicin sulphate USP
Gentamicin sulphate

Aminoglycoside antibiotic. Protein synthesis inhibitor. Causes codon misreading by binding to the 30S ribosomal subunit, blocking the translocation of peptidyl-tRNA from the acceptor site to the donor site. Antibacterial against Gram-negative aerobic bacteria, Gram-positive bacteria and mycoplasmas. Used as a selection agent (gentamicin-resistance gene) in molecular biology applications. Broad-spectrum cell culture antibiotic that is nontoxic to viruses and mammalian cells at antibacterial and antimycoplasmal concentrations. Due to its extended stability and slow development of bacterial resistance, it is a useful antibiotic in long-term virus und tissue culture studies. Bactericidal effects are exerted by the binding to the outer membrane, causing disruption of the membrane. This increases the permeability of the cell envelope, leakage of cell contents, and leading to apoptosis and proteolysis (cell death). Causes also cell death by generation of free radicals, phospholipidosis, extracellular calcium-sensing receptor stimulation and energetic catastrophe.

Formula: C₂₁H₄₅N₅O₁₁S
MDL Number: MFCD00270181
CAS Number: 1405-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR