Okadaic acid sodium salt, high purity

Supplier: ADIPOGEN CORP MS

Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin sodium salt, OA sodium salt

AG-CN2-0062-C025 AG-CN2-0062-C100 AG-CN2-0062-M001
102987-194EA 110.77 USD
102987-194 102987-196 102987-198
Okadaic acid sodium salt, high purity
Okadaic acid sodium salt

Non-phorbol type tumor promoter. Reversible, potent and selective serine threonine protein phosphatase inhibitor. PP2A (IC50=0.2-1nM), PP1 (IC50=3-15nM), PP2B (IC50=>1µM). Does not inhibit PP2C. Stimulates intracellular protein phosphorylation. Useful tool for studying cellular processes that are regulated by phosphorylation. Does not affect activity of acid phosphatase, alkaline phosphatase and tyrosine phosphatase. Mimics the effects of insulin. Activates atypical protein kinase C (zeta/lambda) in 3T3/L1 adipocytes. Enhances transmitter release at neuromuscular junctions. Apoptosis inhibitor. Induces apoptosis in human breast carcinoma cells (MB-231 and MCF-7) and in myeloid cells. Neurotoxic. Used to study various cellular processes including cell cycle, apoptosis, nitric oxide metabolism and calcium signaling. Stimulates cell motility, loss of stabilization of focal adhesions and a consequent loss of cytoskeletal organization.

Formula: C₄₄H₆₇NaO₁₃
MW: 827.00 g/mol
MDL Number: MFCD00210208
CAS Number: 209266-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR