Okadaic acid, high purity

Supplier: ADIPOGEN CORP MS

Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin, OA

AG-CN2-0056-M001 AG-CN2-0056-C025 AG-CN2-0056-C100
102987-174EA 2353.85 USD
102987-174 102987-874 102987-876
Okadaic acid, high purity
Okadaic acid

Non-phorbol type tumor promoter. Reversible, potent and selective serine threonine protein phosphatase inhibitor. PP2A (IC50=0.2-1nM), PP1 (IC50=3-15nM), PP2B (IC50=>1µM). Does not inhibit PP2C. Stimulates intracellular protein phosphorylation. Useful tool for studying cellular processes that are regulated by phosphorylation. Does not affect activity of acid phosphatase, alkaline phosphatase and tyrosine phosphatase. Mimics the effects of insulin. Activates atypical protein kinase C (zeta/lambda) in 3T3/L1 adipocytes. Enhances transmitter release at neuromuscular junctions. Apoptosis inhibitor. Induces apoptosis in human breast carcinoma cells (MB-231 and MCF-7) and in myeloid cells. Neurotoxic. Used to study various cellular processes including cell cycle, apoptosis, nitric oxide metabolism and calcium signaling. Stimulates cell motility, loss of stabilization of focal adhesions and a consequent loss of cytoskeletal organization.

Formula: C₄₄H₆₈O₁₃
MW: 805.02 g/mol
MDL Number: MFCD00083455
CAS Number: 78111-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR