Ingenol 3-angelate

Supplier: ADIPOGEN CORP MS

AG-CN2-0012-M001 AG-CN2-0012-M005
102515-706EA 152.31 USD
102515-706 102987-844
Ingenol 3-angelate
Ingenol-3-angelate

Specific protein kinase C (PKC) activator. Selective activator of PKC isoforms, like PKCtheta in T cells. Antiproliferative and proapoptotic (necrotic). Immunostimulant. Chemotherapeutic. Anticancer compound. Efficacious agent against actinic kerastosis. Antileukemic.

Formula: C₂₅H₃₄O₆
MW: 430.541 g/mol
MDL Number: MFCD07784504
CAS Number: 75567-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR