Octyl-β-D-thioglucopyranoside, Ultrapure

Supplier: ADIPOGEN CORP MS

Synonyms: Octyl-thioglucoside, OTG, 1-S-Octyl-β-D-thioglucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, N-Octyl-1-thio-Beta-D-glucopyranoside, Octyl β-D-thioglucopyranoside

AG-CC1-0005-G001 AG-CC1-0005-G005
102987-828EA 76.15 USD
102987-828 102987-830
Octyl-β-D-thioglucopyranoside, Ultrapure
Octyl β-D-thioglucopyranoside

Non-ionic detergent. For functional solubilization, purification and crystallography of membrane proteins. The thioether linkage makes it resistant to degradation by beta-glucosidase enzymes. For the stabilization and activation of enzymes. Used for cell lysis experiments.

Formula: C₁₄H₂₈O₅S
MW: 308.44 g/mol
MDL Number: MFCD00012189
CAS Number: 85618-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR