Astaxanthine ≥97% (by HPLC)

Supplier: ADIPOGEN CORP MS

AG-CN2-0055-M005 AG-CN2-0055-M025
102987-870EA 69.23 USD
102987-870 102987-872
Astaxanthine ≥97% (by HPLC)
Astaxanthine
Formula: C₄₀H₅₂O₄
MDL Number: MFCD00672621
CAS Number: 472-61-7

Order Now

Specification Test Results

Purity ≥97% (HPLC, NMR)
Appearance Dark violet red solid
Solubility Soluble in DMSO, chloroform or DMF. Soluble in ethanol or methanol if heated. Insoluble in water
Identity Determined by 1H-NMR
Stability Stable for at least 2 years after receipt when stored at -20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR