Green CMFDA ≥95% fluorescent dye, for cytology
Supplier: ADIPOGEN CORP MS
Synonyms:
3',6'-bis(acetyloxy)-5-(chloromethyl)-spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 5-(Chloromethyl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl diacetate, 5-Chloromethylfluorescein diacetate
Thiol-reactive, cell permeant green fluorescent probe with high selectivity.
- Soluble in DMSO
- Purity ≥ 95% (HPLC)
- Well suited for monitoring cell movement or location
- Exhibits ideal tracking properties
Thiol-reactive, cell permeant green fluorescent probe with high selectivity (Ex/Em: 492/517nm). This fluorescent dye is well suited for monitoring cell movement or location. The dye exhibits ideal tracking properties: it is stable, nontoxic at working concentrations, well retained in cells and brightly fluorescent at physiological pH.
Formula:
C₂₅H₁₇ClO₇ MW: 464.9 g/mol |
MDL Number:
MFCD00467170 CAS Number: 136832-63-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...