Dithiothreitol (DTT, Cleland's reagent) ≥99%
Supplier: BeanTown Chemical
Synonyms:
cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol
137105-1G
137105-25G
137105-5G
BT137105-1GEA
38
USD
BT137105-1G
BT137105-25G
BT137105-5G
Dithiothreitol (DTT, Cleland's reagent) ≥99%
Dithiothreitol (DTT, Cleland's reagent)
CAS: 3483-12-3; EC No: 222-468-7; MDL No: MFCD00004877; RTECS: EK1610000
Powder; Molecular Formula: C4H10O2S2; MW: 154.25
Melting Point: 41-44°; Flash point:
- RTECS: EK1610000
- Air Sensitive
- Powder
Formula:
C₄H₁₀O₂S₂ MW: 154.25 g/mol Melting Pt: 41…44 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00004877 CAS Number: 3483-12-3 EINECS: 222-468-7 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...