Puromycin dihydrochloride

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride

0210055210 0210055225
IC10055210EA 53.88 USD
IC10055210 IC10055225
Puromycin dihydrochloride
Puromycin dihydrochloride

Puromycin dihydrochloride is a part of the amino-nucleoside family of antibiotics and is derived from Streptomyces alboniger.


  • Off-white Powder


Puromycin's biological activity are antitumor effect, nephrotoxic action, inhibition of purine and/or protein synthesis and antitrypanosome activity. It inhibits protein synthesis by causing premature chain termination acting as an analog of the 3′-terminal end of the aminoacyl-tRNA. Prevents growth of bacteria, protozoa, algae, and mammalian cells.


Extinction coefficients: 20.3 (275 nm, 0.1 N NaOH), 19.5 (268 nm, 0.1 NHCl) (Lit.)


Soluble in water

Formula: C₂₂H₂₉N₇O₅·2HCl
MW: 544.44 g/mol
Melting Pt: 175.5…177 °C
Storage Temperature: Freezer
MDL Number: MFCD00012691
CAS Number: 58-58-2

Order Now

Specification Test Results

Source Streptomyces albo-niger

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR