Antibiotic PF 1052 (from Phoma sp.) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0338-C500 BVT-0338-M001
102513-774EA 152.31 USD
102513-774 102513-776
Antibiotic PF 1052 (from Phoma sp.) ≥98% (by HPLC)
Antibiotic PF 1052

Antibiotic PF 1052 is antifungal and phytotoxic.


  • Light brown solid


Antibiotic. Antibacterial, antifungal and phytotoxic.


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C


Soluble in methylene chloride, methanol or chloroform.

Formula: C₂₆H₃₉NO₄
MW: 429.6 g/mol
Storage Temperature: Freezer
CAS Number: 147317-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR