5-Hydroxy-2-methyl-4-chromanone ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
BVT-0322-C500
BVT-0322-M001
102513-766EA
173.08
USD
102513-766
102513-768
5-Hydroxy-2-methyl-4-chromanone ≥98% (by HPLC)
5-Hydroxy-2-methyl-4-chromanone
Antifungal and phytotoxic. Inhibits the growth of Saccharomyces cerevisiae and the formation of soybean callus.
Formula:
C₁₀H₁₀O₃ Storage Temperature: Freezer |
CAS Number:
14153-17-4 |
Specification Test Results
Source/Host | Isolated from Nodulisporium sp. |
Purity |
|
Appearance/Color | Off-white to yellow solid. |
Solubility | Soluble in DMSO or methylene chloride; weakly soluble in water. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...