Iromycin A ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

BVT-0262-C500 BVT-0262-M001
102513-682EA 249.23 USD
102513-682 102513-684
Iromycin A ≥98% (by HPLC)
Iromycin A

Pyridone metabolite. Nitric oxide synthase (NOS) inhibitor, showing selectivity for eNOS (NOS III) versus nNOS (NOS I). Thaxtomine biosynthesis inhibitor. Mitochondrial electron transport chain inhibitor.

Formula: C₁₉H₂₉NO₂
Storage Temperature: Freezer
CAS Number: 213137-53-2

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces bottropensis (Gö-Dra17).
Purity 98% (HPLC)
Appearance/Color Off-white to greenish solid.
Solubility Soluble in DMSO; fairly soluble in methanol; poorly soluble in acetone.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR