17-DMAG ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 17-Dimethylaminoethylamino-17-demethoxygeldanamycin

BVT-0255-C100 BVT-0255-M001
102513-660EA 41.54 USD
102513-660 102513-662
17-DMAG ≥98% (by HPLC)
17-DMAG

Less toxic, more potent synthetic derivative of geldanamycin (Prod. No. BVT-0196). Angiogenesis inhibitor. Heat shock protein 90 (HSP90) inhibitor. Apoptosis inducer. Shows higher antitumor activity than 17-AAG (Prod. No. BVT-0244).

Formula: C₃₂H₄₈N₄O₈
MW: 616.8
Storage Temperature: Freezer
MDL Number: MFCD08457919
CAS Number: 467214-20-6

Order Now

Specification Test Results

Source/Host Semi-synthetic derivative from geldanamycin.
Purity 98% (HPLC)
Appearance/Color Purple to black powder.
Solubility Soluble in water, 100% ethanol (10 mg/ml) or DMSO (25 mg/ml)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR