β-Rubromycin (from Streptomyces species) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0251-M001 BVT-0251-M005
102513-640EA 103.85 USD
102513-640 102513-642
β-Rubromycin (from Streptomyces species) ≥98% (by HPLC)
beta-Rubromycin

beta-Rubromycin is an HIV-1 reverse transcriptase inhibitor.


  • Antibiotic
  • Human telomerase inhibitor
  • Cytostatic


Antibiotic. HIV-1 reverse transcriptase inhibitor. Human telomerase inhibitor. Cytostatic against different tumor cell lines.


Dark blue solid


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO or chloroform.

Formula: C₂₇H₂₀O₁₂
MW: 536.4 g/mol
Storage Temperature: Freezer
CAS Number: 27267-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR