Bacimethrin (from Streptomyces albus) ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
(4-Amino-2-methoxypyrimidin-5-yl)methanol
BVT-0226-C500
BVT-0226-M001
102513-620EA
180
USD
102513-620
102513-622
Bacimethrin (from Streptomyces albus) ≥98% (by HPLC)
Bacimethrin
Bacimethrin is a derivative of 5-Hydoxymethylcytosine.
- Antibiotic
- Phosphatase kinase inhibitor
Antibiotic. Antibacterial. Thiamine antagonist. Phosphatase kinase inhibitor.
Off-white powder
After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.
Soluble in DMSO or methanol.
MW:
155.2 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01689392 CAS Number: 3690-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...