Terrein ≥98%

Supplier: ADIPOGEN CORP MS
Warning

BVT-0193-M001 BVT-0193-M005 BVT-0193-M010
102513-586EA 152.31 USD
102513-586 102513-588 102513-590
Terrein ≥98%
Terrein

Antibacterial. Phytotoxin. Melanogenesis inhibitor by reducing tyrosinase production via extracellular signal-regulated protein kinase (ERK) activation. Keratinocyte and epidermis proliferation inhibitor. Mycotoxin. Anti-inflammatory modulator in pulpal inflammation. Angiogenesis inhibitor through angiogenin secretion inhibition. Collagen-induced platelet aggregation inhibitor. Antioxidant agent that enhances osseointegration by decreasing the level of ROS and having a potentially synergistic effect on osteoblast differentiation. Proteasome inhibitor.

Formula: C₈H₁₀O₃
Storage Temperature: Freezer
CAS Number: 582-46-7

Order Now

Specification Test Results

Source/Host Isolated from Aspergillus terreus.
Purity 98% (HPLC).
Appearance/Color White to off-white solid.
Solubility Soluble in methanol or DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR