Antibiotic UK-1 (from Streptomyces sp. K17/9) ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
BVT-0013-C250
BVT-0013-M001
102513-458EA
110.77
USD
102513-458
102513-460
Antibiotic UK-1 (from Streptomyces sp. K17/9) ≥98% (by HPLC)
Antibiotic UK-1
Antibiotic UK-1 is a potent anti-cancer compound.
- Antibiotic
- Topoisomerase II (Topo II) inhibitor
- Anti-cancer compound
Antibiotic. Antifungal. Topoisomerase II (Topo II) inhibitor. Mg2+- and Zn2+-dependent, DNA binding agent. Potent anti-cancer compound.
White to off-white solid
After reconstitution protect from light at -20°C. Stable for at least 1 year after receipt when stored at -20°C.
Formula:
C₂₂H₁₄N₂O₅ MW: 386.4 g/mol Storage Temperature: Freezer |
CAS Number:
151271-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...