γ-Rubromycin (from Streptomyces species) ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0007-M001 BVT-0007-M005
102513-450EA 110.77 USD
102513-450 102513-452
γ-Rubromycin (from Streptomyces species) ≥98% (by HPLC)
gamma-Rubromycin

gamma-Rubromycin is a HIV-1 reverse transcriptase and human telomerase inhibitor.


  • Antibiotic
  • Active against different tumor cell lines
  • Anti-bacterial


Antibiotic. HIV-1 reverse transcriptase and human telomerase inhibitor. Antibacterial. Cytostatically active against different tumor cell lines.


Red powder


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in methylene chloride, DMSO or methanol.

Formula: C₂₆H₁₈O₁₂
MW: 522.4 g/mol
Storage Temperature: Freezer
CAS Number: 27267-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR