Rosiglitazone maleate ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: 5-(4-(2-(Methyl(pyridin-2-yl)amino)ethoxy)benzyl)thiazolidine-2,4-dione maleate

AG-CR1-3571-M025 AG-CR1-3571-M100 AG-CR1-3571-G001
102516-324EA 69.23 USD
102516-324 102516-326 102516-328
Rosiglitazone maleate ≥98%
Rosiglitazone maleate

Same activities as rosaglitazone (Prod. No. AG-CR1-3570) but different formulation. Antidiabetic, hypoglycemic agent. Potent and selective peroxisome proliferator-activated receptor gamma (PPAR-gamma) agonist. Potent insulin sensitizing agent binding to the PPAR receptors in fat cells and making the cells more responsive to insulin. Ameliorates insulin resistance. Improves blood pressure and vascular function. Enhances proliferation of endogenous neural progenitor cells (NPCs). Anti-inflammatory compound. Has controversial therapeutic effects on the cardiovascular system. Promotes adipocyte differentiation of mesenchymal stem cells (MSCs).

Formula: C₁₈H₁₉N₃O₃S·C₄H₄O₄
MW: 473.4989 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03427306
CAS Number: 155141-29-0

Order Now

Specification Test Results

Purity 98% (HPLC)
Appearance/Color White to off-white powder.
Solubility Soluble in ethanol or buffered aqueous solutions with pH of 2.3.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR