S14161 ≥99% (by NMR)
Supplier: ADIPOGEN CORP MS
AG-CR1-3516-M001
AG-CR1-3516-M005
102516-268EA
80.31
USD
102516-268
102516-270
S14161 ≥99% (by NMR)
S14161
Potent antileukemia and antimyeloma compound. Apoptosis inducer. Delays tumor growth. Cyclin D2 promoter transactivation inhibitor. Inhibitor of pan-Class I PI3K isoforms.
Formula:
C₁₇H₁₄FNO₄ Storage Temperature: Freezer |
CAS Number:
883046-50-2 |
Specification Test Results
Purity |
|
Appearance/Color | Yellow solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...