Kaempferitrin ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 5-Hydroxy-2-(4-hydroxyphenyl)-3,7-bis(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

AG-CN2-0039-M001 AG-CN2-0039-M005
102515-814EA 69.23 USD
102515-814 102515-816
Kaempferitrin ≥98% (by NMR)
Kaempferitrin

Cytotoxic and anti-inflammatory compound (weak). Antioxidant. Lipid peroxidation inhibitor. Free radical scavenger. Inhibitor of myeloperoxidase activity. Hypoglycemic. Decreases blood glucose levels in diabetic and normal rats. Insulinomimetic. Modulates glucose homeostasis. Decreases blood glucose levels and stimulates glucose uptake in muscle. Inhibits GLUT4 mediated glucose uptake in differentiated 3T3-L1 cells. Improves insulin resistance by the activation of the classical insulin transduction pathway. Increases adiponectin secretion in 3T3-L1 adipocytes. Antinoniceptive. May have antidyslipidemic activity.

Formula: C₂₇H₃₀O₁₄
MW: 578.5
Storage Temperature: Freezer
MDL Number: MFCD01662671
CAS Number: 482-38-2

Order Now

Specification Test Results

Source/Host Isolated from Calendula officinalis.
Purity 98% (NMR)
Appearance/Color Yellow solid.
Solubility Soluble in ethanol or DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR