Troglitazone ≥98%
Supplier: ADIPOGEN CORP MS
AG-CR1-3565-M005
AG-CR1-3565-M025
102516-294EA
76.15
USD
102516-294
102516-296
Troglitazone ≥98%
Troglitazone
Potent and selective PPARgamma agonist with no activation of PPARalpha and PPARdelta. Anti-diabetic agent with potent glucose-lowering effects and decreasing insulin resistance in adipose tissues. Stimulates adipogenesis. Apoptosis and cell cycle arrest inducer in several cancer cell lines. Anti-inflammatory. Inhibits vascular smooth muscle cell (VSMC) migration.
Formula:
C₂₄H₂₇NO₅S Storage Temperature: Freezer |
MDL Number:
MFCD00878416 CAS Number: 97322-87-7 |
Specification Test Results
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in DMSO, ethanol or dimethylformamide (DMF). |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...