PP2 ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 1-(tert-Butyl)-3-(4-chlorophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine

AG-CR1-3563-M001 AG-CR1-3563-M005
102516-284EA 55.38 USD
102516-284 102516-286
PP2 ≥98% (by HPLC)
PP2

Highly potent and selective Scr family tyrosine kinase inhibitor. Reduces cancer metastasis. Apoptotic. Antitumor compound. RIP2 inhbitor. Blocks TGF-beta-mediated cellular responses. Autophagy modulator.

Formula: C₁₅H₁₆ClN₅
MW: 301.8 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01568210
CAS Number: 172889-27-9

Order Now

Specification Test Results

Purity 98% (HPLC)
Appearance/Color White solid.
Solubility Soluble in DMSO. Slightly soluble in ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR