GANT61 ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: 2,2'-((2-(Pyridin-4-yl)dihydropyrimidine-1,3(2H,4H)-diyl)bis(methylene))bis(N,N-dimethylaniline)

AG-CR1-3561-M001 AG-CR1-3561-M005
102516-276EA 55.38 USD
102516-276 102516-278
GANT61 ≥98%
GANT61

Cell permeable GLI antagonist. Inhibits Gli1 and Gli2-induced transcription. Sonic hedgehog (Shh) signaling pathway inhibitor, downstream of SMO and SUFU. Displays antiproliferative and antitumor activity in vitro and in vivo. Apoptosis inducer.

Formula: C₂₇H₃₅N₅
MW: 429.6 g/mol
Storage Temperature: Freezer
MDL Number: MFCD14635408
CAS Number: 500579-04-4

Order Now

Specification Test Results

Purity 98%
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR