SANT-2 ≥95% (by NMR)
Supplier: ADIPOGEN CORP MS
Synonyms:
N-(3-(1H-Benzo[d]imidazol-2-yl)-4-chlorophenyl)-3,4,5-triethoxybenzamide
AG-CR1-3514-M001
AG-CR1-3514-M005
102516-260EA
48.46
USD
102516-260
102516-262
SANT-2 ≥95% (by NMR)
SANT-2
Sonic hedgehog (Shh) signaling pathway modulator. Cell permeable potent smoothened (Smo) antagonist. Inhibits wt and oncogenic Smo. SAG activity (Prod. Nr. AG-CR1-3506) inhibitor. Gli1 reporter gene expression inhibitor. Suppresses the Shh-dependent translocation of Smo to the primary cilium. Potential tumor growth blocker.
Formula:
C₂₆H₂₆ClN₃O₄ MW: 480 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01827636 CAS Number: 329196-48-7 |
Specification Test Results
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in DMSO, ethanol or chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...