C646 ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 4-(4-((5-(4,5-Dimethyl-2-nitrophenyl)furan-2-yl)methylene)-3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzoic acid

AG-CR1-3508-M001 AG-CR1-3508-M005
102516-238EA 48.46 USD
102516-238 102516-240
C646 ≥98% (by NMR)
C646

Reversible cell permeable p300/CBP histone acetyltransferase (HAT) inhibitor. Specific inhibition to p300 (86%) compared to N-acetyltransferase, PCAF, GCN5, Rtt109, Sas or MOZ histone acetyltransferases (10%). Cell growth inhibitor in melanoma and non-small-cell-lung (NSCL) human cancer cell lines.

Formula: C₂₄H₁₉N₃O₆
MW: 445.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01784780
CAS Number: 328968-36-1

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Red solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR