TPh A ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

AG-CR1-3507-M001 AG-CR1-3507-M005
102516-234EA 48.46 USD
102516-234 102516-236
TPh A ≥98% (by NMR)
TPh A

Nuclear protein pirin inhibitor. Inhibits the interaction between Bcl3 oncoprotein and the nuclear protein pirin. Melanoma cell migration inhibitor.

Formula: C₂₁H₂₁NO₃S₂
Storage Temperature: Refrigerator
CAS Number: 21306-65-0

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR