Ds(+)-threo-Isocitric acid trisodium salt ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

AG-CR1-3505-M025 AG-CR1-3505-M100
102516-226EA 76.15 USD
102516-226 102516-228
Ds(+)-threo-Isocitric acid trisodium salt ≥98% (by NMR)
Ds(+)-threo-Isocitric acid trisodium salt

Anticoagulant that retains platelets functionality. Buffers Ca2+ activity in physiological range (~1mM). Bypasses the metabolic blockade, may permit heme biosynthesis but does not suffice to reverse the iron-restriction response. Stabilizes the iron-sulfur cluster and is predicted to restore aconitase to its 'high iron' conformation and thereby dissociates the PKC-associated signaling complexes whose hyperactivation impairs Epo responsiveness. Binds IDH (isocitrate dehydrogenase), an excellent candidate for participating, along with aconitase and PKC, in an iron restriction signalosome. Contributes to heme biosynthesis which may account for its greater potency in restoring erythroid differentiation.

Formula: C₆H₅O₇·Na
Storage Temperature: Freezer
CAS Number: 903507-52-8

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White solid.
Solubility Soluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR