3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione ≥98% (by NMR)
Supplier: ADIPOGEN CORP MS
Synonyms:
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione, 3,4-Bis(3-indolyl)maleimide, Bisindolylmaleimide IV, Arcyriarubin A
AG-CR1-0152-M001
AG-CR1-0152-M005
102516-208EA
76.15
USD
102516-208
102516-210
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione ≥98% (by NMR)
3,4-Di(1H-indol-3-yl)-1H-pyrrole-2,5-dione
Cell permeable protein kinase C (PKC) inhibitor. Inhibits also protein kinase A (PKA).
Formula:
C20H13N3O2 MW: 327.3361 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00236432 CAS Number: 119139-23-0 |
Specification Test Results
Purity |
|
Appearance/Color | Red to orange solid. |
Solubility | Soluble in DMSO, ethanol, methanol or dimethylformamide. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...