MnTBAP chloride ≥97%

Supplier: ADIPOGEN CORP MS

Synonyms: MnTBAP, [5,10,15,20-Tetrakis(4-carboxyphenyl)-21H,23H-porphine]manganese(III) chloride

AG-CR1-0133-M010 AG-CR1-0133-M050
102516-194EA 34.62 USD
102516-194 102516-196
MnTBAP chloride ≥97%
MnTBAP chloride

Cell permeable superoxide dismutase (SOD) mimetic. Potent inhibitor of peroxynitrite-induced oxidative reactions (peroxynitrite scavenger), but not a scavenger of nitric oxide (NO). Neuronal apoptosis inhibitor. Protects T cells from superoxide generation, caspase-dependent DNA loss and cell death. Lipopolysaccharide-induced TNF-alpha production inhibitor, by prevention of intracellular ROS generation and subsequent inactivation of p38 MAPK and SAPK/JNK. .

Formula: C₄₈H₂₈MnN₄O₈Cl
MW: 879.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD06227760
CAS Number: 55266-18-7

Order Now

Specification Test Results

Purity 97%
Appearance/Color Black crystalline powder.
Solubility Dissolve at 0.1M in aqueous base, then add buffer to lower pH to pH 7.0 or higher as needed.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR