YC-1 ≥98% (by NMR)
Supplier: ADIPOGEN CORP MS
Nitric oxide (NO) independent, superoxide-sensitive soluble guanylyl cyclase (sGC) activator. Induces concentration-dependent increase in cGMP levels. Inhibits platelet adhesion to collagen. Thombosis inhibitor. Non-specific phosphodiesterase inhibitor. Na+ channel blocker. Anticancer compound. HIF-alpha inhibitor. Blocks angiogenesis. Tumor growth inhibitor. Apoptosis inducer. NK cell differentiation enhancer. TRAIL sensitizer.
Formula:
C₁₉H₁₆N₂O₂ Storage Temperature: Freezer |
MDL Number:
MFCD06407798 CAS Number: 170632-47-0 |
Specification Test Results
Purity |
|
Appearance/Color | White to pale yellow powder. |
Solubility | Soluble in DMSO, methanol or dichloromethane. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...