GW1929 ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

Synonyms: N-(2-Benzoylphenyl)-L-tyrosine PPARγ agonist

AG-CR1-0116-M001 AG-CR1-0116-M005 AG-CR1-0116-M025
102516-174EA 41.54 USD
102516-174 102516-176 102516-178
GW1929 ≥98% (by NMR)
GW1929

Potent and subtype-selective (1'000-fold) PPARgamma agonist. Angiogenesis inhibitor. Apoptosis inhibitor. Anti-inflammatory. Anti-hyperglycemic and anti-hyperlipidemic agent. Antidiabetic. The glucose-lowering effect in rats is 100-fold more potent than that of troglitazone.

Formula: C₃₀H₂₉N₃O₄
MW: 495.579 g/mol
Storage Temperature: Freezer
CAS Number: 196808-24-9

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Off-white to yellow crystalline solid.
Solubility Soluble in DMSO or methanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR