Bisindolylmaleimide VIII·acetate ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

AG-CR1-0114-M001 AG-CR1-0114-M005
102516-170EA 62.31 USD
102516-170 102516-172
Bisindolylmaleimide VIII·acetate ≥98% (by NMR)
Bisindolylmaleimide VIII acetate

Selective protein kinase C (PKC) inhibitor. Inhibits proliferation of cultured mouse epidermal keratinocytes. Enhances Fas- and TRAIL-mediated apoptosis. Inhibits T cell-mediated autoimmune diseases. Inhibits stimulation of insulin secretion by glucose. Inhibits panel of protein kinases (e.g. GSK-3beta, CDK2).

Formula: C₂₄H₂₂N₄O₂·CH₃COOH
Storage Temperature: Freezer
MDL Number: MFCD00909467
CAS Number: 138516-31-1

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Red powder.
Solubility Soluble in DMSO, methanol or water. Slightly soluble in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR