Bisindolylmaleimide III ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

AG-CR1-0112-M001 AG-CR1-0112-M005
102516-162EA 76.15 USD
102516-162 102516-164
Bisindolylmaleimide III ≥98% (by NMR)
Bisindolylmaleimide III

Potent and selective protein kinase C (PKC) inhibitor. Inhibits panel of protein kinases (e.g. GSK-3beta, CDK2).

Formula: C₂₃H₂₀N₄O₂
Storage Temperature: Freezer
MDL Number: MFCD01076633
CAS Number: 137592-43-9

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Red solid.
Solubility Soluble in methanol or DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR