Bisindolylmaleimide I·hydrochloride ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

AG-CR1-0110-M001 AG-CR1-0110-M005
102516-154EA 76.15 USD
102516-154 102516-156
Bisindolylmaleimide I·hydrochloride ≥98% (by NMR)
Bisindolylmaleimide I hydrochloride

Cell permeable kinase inhibitor with improved selectivity for protein kinase C (PKC) (Ki = 10 nM). Competitive inhibitor for the ATP-binding site of PKC. Anti-inflammatory. Binds to P-glycoprotein. Telomerase activity inhibitor. Potent glycogen synthase kinase-3 (GSK-3) inhibitor. Necrosis inhibitor. Blocks hERG potassium channels. Promotes osteoblastogenesis in human mesenchymal stem cells.

Formula: C₂₅H₂₄N₄O₂·HCl
MW: 452.5·36.5 g/mol
Storage Temperature: Freezer
CAS Number: 176504-36-2

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Orange to red powder.
Solubility Soluble in DMSO or methanol. Slightly soluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR