Bisindolylmaleimide XI·hydrochloride ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
AG-CR1-0109-M001
AG-CR1-0109-M005
102516-150EA
117.69
USD
102516-150
102516-152
Bisindolylmaleimide XI·hydrochloride ≥98% (by HPLC)
Bisindolylmaleimide XI hydrochloride
Selective cell permeable protein kinase C (PKC) inhibitor. G protein-coupled receptor kinase (GRK-5) inhibitor. Less potent GRK-2 or GRK-3 inhibitor. Prevents T cell-driven chronic inflammatory responses in vivo.
Formula:
C₂₈H₂₈N₄O₂·HCl MW: 452.5·36.5 g/mol Storage Temperature: Freezer |
CAS Number:
145333-02-4 |
Specification Test Results
Purity |
|
Appearance/Color | Dark red solid. |
Solubility | Soluble in DMSO or methanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...