PJ-34 ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

Synonyms: 2-(Dimethylamino)-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride, PJ34 HCl

AG-CR1-0100-M001 AG-CR1-0100-M005 AG-CR1-0100-M025
102516-138EA 55.38 USD
102516-138 102516-140 102516-142
PJ-34 ≥98% (by NMR)
PJ-34 hydrochloride

Potent, water soluble poly(ADP-ribose) polymerase (PARP) inhibitor. Inhibits peroxynitrite-induced cell necrosis. Shows significant, dose-dependent anti-inflammatory effects in a variety of local inflammation models. Provides cardioprotection by decreasing myocardial infarct size. Shows protective effects in models of stroke. Suppresses cell growth in liver cancer cells.

Formula: C₁₇H₁₇N₃O₂·HCl
MW: 295.2·36.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22581510
CAS Number: 344458-15-7

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to light brown powder.
Solubility Soluble in DMSO or water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR