Pioglitazone ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 5-(4-(2-(5-Ethylpyridin-2-yl)ethoxy)benzyl)thiazolidine-2,4-dione

AG-CR1-0067-M001 AG-CR1-0067-M005 AG-CR1-0067-M025
102516-118EA 27.69 USD
102516-118 102516-120 102516-122
Pioglitazone ≥98%
Pioglitazone

Thiazolidinedione (TZD) reference compound. Selective PPARgamma agonist. Selectively activates PPARgamma-1. Anti-diabetic , decreasing insulin resistance in adipose tissue. Is about one tenth as potent as rosiglitazone. Lipid metabolism modulator. Estrogen synthesis inhibitor. NF-kappaB inhibitor. Neuroprotective.

Formula: C₁₉H₂₀N₂O₃S
Storage Temperature: Freezer
MDL Number: MFCD00865504
CAS Number: 111025-46-8

Order Now

Specification Test Results

Purity 98% (1H-NMR).
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO (2.5 mg/ml) or dimethylformamide (DMF) (2.5 mg/ml).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR