PD 150,606 ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Danger

AG-CR1-0066-M005 AG-CR1-0066-M025
102516-114EA 96.92 USD
102516-114 102516-116
PD 150,606 ≥98% (by NMR)
PD 150,606

Cell permeable and selective calpain-1 and -2 inhibitor. Interacts with calcium-binding domain of calpain. Neuroprotective. Ca2+-permeable AMPA receptor inhibitor. Apoptosis inhibitor Autophagy activator by preventing cleavage of ATG5

Formula: C₉H₇IO₂S
Storage Temperature: Freezer
MDL Number: MFCD00949076
CAS Number: 426821-41-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purity >98% (NMR)
Appearance/Color Off-white solid.
Solubility Soluble in DMSO or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR